Learn More
PureAire Monitoring Systems, Inc. Remote Digital LED Display for Monitors
The Remote Digital Display provides continuous information on the status of the PureAire Oxygen Deficiency Monitor by remotely mirroring the monitor’s display and alarm panel. A large, bright red LED display continuously illuminates the O2 reading from the oxygen monitor up to 75 meters.
Brand:  PureAire Monitoring Systems, Inc. 99091
Product Code. 16268821
  • 590.00 € / Each
Explore more special offers
Description
Description
  • Bright 1 in. red LED digital concentration display
  • Built-in alarm and fault status LED’s
  • User selectable dual level alarm set points
  • Built-in audio horn alerts personnel to low or high oxygen conditions
  • Two tone horn distinguishes between alarm 1 and alarm 2
  • Displays Oxygen monitor overall status
  • The Remote Digital Alarm Indicator provides continuous information on the status of the PureAire oxygen deficiency monitors or analysers by remotely mirroring the monitor’s display. A large, bright red LED display continuously illuminates the O2 reading from the oxygen monitor up to 250 feet away.
  • You can select two individual alarm set points to activate the visual alarm LED’s and audible horn. High intensity LED’s illuminate whenever preset alarm levels are exceeded. Also included is an additional fault LED indicator for system errors. The remote digital alarm indicator has a 90dB built-in horn to alert personnel when an unsafe condition exists.
  • It connects to an oxygen monitor using a 3-wire, 18 gauge shielded cable. The remote display can also be connected to a Programmable Logic Controller (PLC), providing simultaneous alarm and status indications in multiple locations throughout the facility.
Services
2 years
Specifications
Specifications
Horn Sound Output 80 – 85 db
3.5 in.
2 in.
-40°F to +122°F
24 VDC
1.1 lb.
4.6 in.
2 x 4.6 x 3.5 in.
88.9 mm
50.8 mm
-40 to +50°C
2 Year after ETD
0.5 kg
116.84 mm