Learn More
Fisherbrand™ Remote Dispenser with Sensor
Includes a built-in high precision resistivity sensor with cell constant of 0.01 cm-1. Fisher Scientific™ Remote Dispenser with Sensor can measure the quality of ultrapure water just prior to the dispensing point.
Brand:  Fisherbrand™ RSP0U000S
Product Code. 15419759
  • 1545.00 € / Each
Estimated Shipment: 12-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Point of Use remote water dispenser
  • Built-in high precision resistivity sensor with cell constant of 0.01cm-1 to measure the quality of ultrapure water just prior to the dispensing point
  • Volumetric dispensing and calibration
  • 360 degree-rotatable and detachable handle
  • Effortless and instant dispensing with maximum flexibility
Specifications
Specifications
Accu500
Remote Water Dispenser