Learn More
Thermo Scientific™ Repair Kits for Thermo Scientific™ Plunger-in-Needle Syringes
Extend the life and performance, and reduce the cost per use of your syringes with the Thermo Scientific™ Repair Kit for Thermo Scientific Plunger-in-Needle Syringes.
59.75 €
Specifications
Needle Gauge | 23 G |
---|---|
Product Type | Syringe Repair Kit |
Connection Type | Plunger-In |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
10017684
|
Thermo Scientific™
365RK341 |
PIN Syringe 365B0531 | N/A | N/A | N/A | |||||
Description
Thermo Scientific™ Repair Kits for Thermo Scientific Plunger-in-Needle Syringes extend the life and performance and reduce cost per use of syringes.
- Repair kits for plunger in needle syringes contain both the plunger and needle, which must be changed together
Includes: Plunger, 1; Needle, 1
Specifications
23 G | |
Syringe Repair Kit | |
Plunger-In |