Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile
GREENER_CHOICE
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
Brand:  Corning™ 6632
 View more versions of this product
Product Code. 15863905
  • 96.06 € / Pack of 100
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
  • Pyrogen-, ATP-, RNase-, and DNase-free
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Polypropylene
Dispensing of viscous and volatile liquids
5 mL
ISO8655 ISO11137
Sterile
100/Pk.