Learn More
UVP Replacement Bulbs
Brand:  UVP 34-0014-01
 View more versions of this product
Product Code. 12822969
  • 11.15 € / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Replacement Bulb
6 W
9.5 x 2.8 in. (24.1 x 7.1 cm)
Replacement 6 W tube
24.1 cm
2.8 in.
115 V
365/254 nm
Models UVGL-48, ML-49 and UVG-47 UV lamp
9.5 in.
1 Ea.
7.1 cm