Learn More
UVP Replacement Bulbs
Brand:  UVP 34-0002-01
 View more versions of this product
Product Code. 12863848
  • 34.70 € / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
115 V
Models C-70G, C-71 chromato-vue cabinet
1 Ea.
25 W
Replacement 25 W tube/bulb
Replacement Bulb