Learn More
Thermo Scientific™ Replacement Microcentrifuge Rotor Lids
Combine these replacement microcentrifuge rotor lids with the microcentrifuge rotor.
362.81 € - 373.86 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10554573
|
Thermo Scientific™
75003406 |
Screw-on Lid |
362.81 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10177222
|
Thermo Scientific™
75003410 |
ClickSeal™ Biocontainment Lid |
373.86 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Combine Thermo Scientific™ Replacement Microcentrifuge Rotor Lids with the microcentrifuge rotor.
ClickSeal™ Biocontainment Lid:
- For use with 24 × 1.5/2.0 mL Rotor (75003424), PCR 4 × 8 Rotor (75003440)
- Unique design clicks when proper seal is made, providing assurance that your samples are securely contained, as well as shortens retrieval time with one click opening
- With PPSU rotor lid, view contents of rotor for added safety and convenience
Screw-on Lid:
- For use with 36 × 0.5 mL Rotor (75003436), Dual Row 18 × 2.00.5 mL Rotor (75003418), PCR 8 × 8 Rotor (75003489)
Ordering Information:
Sold individually.