missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Replacement Needles for LC Syringes

Thermo Scientific™ Replacement Needles for LC Syringes are available for syringes with removable needles.

Specifications

Needle Gauge 22 G
Length (English) Needle 2 in.
Product Type Syringe Needle
Length (Metric) Needle 50.8 mm
For Use With (Equipment) LC Autosampler Syringe
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10111083
View Documents Promotion Details
Thermo Scientific™
365RNL15
10 μL N/A
N/A
10079142
View Documents Promotion Details
Thermo Scientific™
365RNL25
25 to 500 μL N/A
N/A
Description

Description

Thermo Scientific™ Replacement Needles for LC Syringes are available for syringes with removable needles.

Specifications

Specifications

22 G
Syringe Needle
LC Autosampler Syringe
2 in.
50.8 mm
Removable
Videos
SDS
Documents

Documents

Product Certifications