Learn More
Fisherbrand™ Replacement Parts for MultiSUB Maxi Horizontal Gel System
Spare safety lid and electrophoresis tank for use with 20 x 10cm, 20 x 15cm, 20 x 20cm or 20 x 25cm gel trays. Fisherbrand™ Replacement Parts for MultiSUB Maxi Horizontal Gel System are constructed of high-quality, injection molded plastic for use in extended electrophoresis separations.
Brand:  Fisherbrand™ MS20TANK
 View more versions of this product
Product Code. 11803473
  • 346.00 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
MultiSUB Horizontal Gel Electrophoresis Systems combine economical buffer and gel consumption without compromising resolution and separation speed.
The large format of the MultiSUB Maxi enables the rapid separation of up to 550 samples. The MS Maxi is versatile enough for routine preparatory and high-throughput techniques, including RFLP analysis, southern and northern blotting applications, and DNA library and PCR fragment analysis. It is primarily designed for separating high numbers of samples from PCR or cloning.
  • Electrophoresis Tank (Including Electrodes): A raised platform holds the UV sample trays, with side wells to hold buffer solution; connects to an external power source
  • Asymmetrical Safety Lid: Notes the correct polarity and orientation of electrodes, with guides that enable the electrode thumb locators to click easily into the correct position
  • Tank and lid are designed for use with 20 x 10cm, 20 x 15cm, 20 x 20cm or 20 x 25cm gel trays
Specifications
Specifications
Clear
MultiSUB Maxi Horizontal Gel Electrophoresis Unit
20 x 10 cm, 20 x 15 cm, 20 x 20 cm or 20 x 25 cm
For up to 550 separations
Nucleic Acid Gel Electrophoresis
Tank with Electrodes
Molded Plastic
1 Each