Learn More
Fisherbrand™ Replacement Parts for MultiSUB Midi Horizontal Gel System
Spare safety lid and electrophoresis tank for use with 10 x 7cm or 10 x 10cm gel trays. Fisherbrand™ Replacement Parts for MultiSUB Midi Horizontal Gel System are constructed of high-quality, injection molded plastic for use in nucleic acid gel electrophoresis applications.
Brand:  Fisherbrand™ MS10TANK
 View more versions of this product
Product Code. 11853353
  • 189.00 € / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
MultiSUB Horizontal Gel Electrophoresis Systems combine economical buffer and gel consumption without compromising resolution and separation speed.
The MultiSUB Midi is has a wider format than the Mini and is the preferred option for quick sample checks of medium volumes, particularly during PCR and cloning. It is an economical choice for rapid separation of up to 100 samples.
  • Electrophoresis Tank (Including Electrodes): A raised platform holds the UV sample trays, with side wells to hold buffer solution; connects to an external power source
  • Asymmetrical Safety Lid: Notes the correct polarity and orientation of electrodes, with guides that enable the electrode thumb locators to click easily into the correct position
  • Tank and lid are designed for use with both 10 x 7cm or 10 x 10cm gel trays
Specifications
Specifications
Clear
MultiSUB Midi Horizontal Gel Electrophoresis Unit
10 x 7 cm or 10 x 10 cm
For up to 100 separations
Nucleic Acid Gel Electrophoresis
Tank with Electrodes
Molded Plastic
1 Each