Learn More
Scientific Industries SI™ Rubber Cover
Fits on 76.2mm Replacement Platform. Scientific Industries SI™ Rubber Cover is used with Vortex-Geinie 1 and Vortex-Genie 2 family of mixers and Vortex-Genie Pulse.
Brand: Scientific Industries SI™ 580-2013-00
This item is not returnable.
View return policy
Description
- For use with Vortex-Genie family of products
- Platform attachment required
Specifications
Rubber | |
Vortex-Genie 1 and Vortex-Genie 2 family of mixers, Vortex-Genie Pulse | |
Replacement Platform 15547315 |