Learn More
Grant Instruments™ Polycarbonate Transparent Lid, Gabled
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
Brand:  Grant Instruments™ AQL26
 View more versions of this product
Product Code. 11406427
  • 193.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
  • Polycarbonate construction
  • Clear, gabled lid allows condensation buildup to drip away from immersed vessels.
  • For use with Sub Aqua Pro Series Unstirred Water Baths
Specifications
Specifications
Gabled
26L Tanks
Polycarbonate