Learn More
Grant Instruments™ Polycarbonate Transparent Lid, Gabled
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
Brand:  Grant Instruments™ AQL5
 View more versions of this product
Product Code. 11486417
  • 119.00 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
  • Polycarbonate construction
  • Clear, gabled lid allows condensation buildup to drip away from immersed vessels.
  • For use with Sub Aqua Pro Series Unstirred Water Baths
Specifications
Specifications
Gabled
5L Tanks. Fits the Grant JBA5 bath, and would be a valid replacement lid for the JBN5.
Polycarbonate