Learn More
Masterflex™ C/L™ Power supply for VDC Powered Variable-Speed Pumps; 100 to 240 VAC
Safely adapt your VDC powered C/L™ pump
Brand:  Masterflex™ 77120-11
Product Code. 13408106
  • 196.00 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Suitable for use with C/L™ pumps
Order Info
Voltage
Specifications
Specifications
Replacement Power Supply Accessory
115/230 V