Learn More
Trajan Scientific and Medical™ Replacement SilTite™ Mini Union Ferrules
Delivers an easy, leak-free installation for capillary columns without the use of any tools.
Brand: Trajan Scientific and Medical™ 073470
This item is not returnable.
View return policy
Description
- Specifically designed for connecting capillary columns
- Connects guard columns to capillary columns
- Inert and precise glass lined bore
- Low dead volume
Specifications
SilTite Mini Union Ferrules for 0.1 to 0.25 mm ID Columns | |
0.1 to 0.25 mm | |
For 0.1 to 0.25 mm Columns | |
10/Pk. |