Learn More
Scienceware Mercury Collector Jar with Integrated Sponge and Perforated Plate
The Mercury Collector Jar with Integrated Sponge and Perforated Plate makes small mercury spill clean-up quick and simple.
Brand:  Scienceware 06706-00
Product Code. 13064897
  • 46.10 € / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Foam pad on lid liner absorbs spilled mercury.
  • Simply screw on lid to collect mercury beneath perforated plate.
Specifications
Specifications
Mercury Spill Kit
4 in.
2 in.
Mercury Collection
10.16 cm
5.08 cm