Learn More
Cole-Parmer™ Replacement UV Tubes
Use these replacement UV tubes with ultraviolet lamps.
Brand:  Cole-Parmer™ 09815-55
Product Code. 11830250
  • 89.10 € / Each
Estimated Shipment: 27-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Replacement Ultraviolet Tubes
  • Product may contain trace amounts of mercury
  • Use caution with breakage, cleanup, shipping and/or disposal
Specifications
Specifications
15 W
365 nm
25.5 mm
UV Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Replacement UV Tubes > Wattage: 15w; Wavelength: 365nm; For deluxe ultraviolet viewing cabinet