Learn More
Brady™ Repositionable Die-cut Vinyl Cloth Label Tape for M6/M7 Printers
Once applied, CleanLift™ Series Vinyl Cloth Labels are permanent or repositionable.
42.20 € - 104.00 €
Specifications
Autoclavable | Not Autoclavable |
---|---|
Adhesive Type | Repositionable Rubber |
Coating | None |
Color | White |
Finish | Semi-gloss |
Product Code | Brand | Certifications/Compliance | Dimensions (LxW) | For Use With (Equipment) | Height (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Certifications/Compliance | Dimensions (LxW) | For Use With (Equipment) | Height (Metric) | Price | Quantity & Availability | ||
17818304
|
Brady™
173312 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 0.5 x 0.5 in. | M710, BMP71 | 12.7 mm |
70.10 €
Pack of 500 |
In Stock
Log in to see stock. |
||
17898244
|
Brady™
174348 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 0.5 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 12.7 mm |
74.85 €
Pack of 500 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17818814
|
Brady™
173405 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 0.5 x 1 in. | M710, BMP71 | 12.7 mm |
80.55 €
Pack of 500 |
In Stock
Log in to see stock. |
||
17808464
|
Brady™
174214 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 0.5 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 12.7 mm |
86.15 €
Pack of 500 |
In Stock
Log in to see stock. |
||
17819104
|
Brady™
173356 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 0.5 x 1.5 in. | M710, BMP71 | 12.7 mm |
71.20 €
Pack of 500 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17889164
|
Brady™
174264 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 0.5 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 12.7 mm |
76.15 €
Pack of 500 |
In Stock
Log in to see stock. |
||
17809024
|
Brady™
173428 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 0.75 x 0.5 in. | M710, BMP71 | 19.05 mm |
64.00 €
Pack of 500 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17888214
|
Brady™
174183 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 0.75 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 19.05 mm |
68.40 €
Pack of 500 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17848194
|
Brady™
174176 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 0.75 x 0.25 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 19.05 mm |
78.55 €
Pack of 750 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17899024
|
Brady™
173431 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 0.75 x 0.25 in. | M710, BMP71 | 19.05 mm |
73.45 €
Pack of 750 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17858934
|
Brady™
174554 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 1 x 0.375 in. | M710, BMP71 | 25.4 mm |
76.50 €
Pack of 500 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17891669
|
Brady™
173720 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 1 x 0.375 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm |
71.75 €
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17828464
|
Brady™
174217 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 1 x 0.75 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm |
56.55 €
Pack of 250 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||
17859174
|
Brady™
173402 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 1 x 0.75 in. | M710, BMP71 | 25.4 mm |
52.85 €
Pack of 250 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17849134
|
Brady™
173396 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 1 x 1 in. | M710, BMP71 | 25.4 mm |
57.20 €
Pack of 250 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17898264
|
Brady™
174224 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 1 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm |
61.10 €
Pack of 250 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17888994
|
Brady™
174279 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 1 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm |
77.85 €
Pack of 250 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17851419
|
Brady™
173843 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 1.5 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 38.1 mm |
84.50 €
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17858794
|
Brady™
173348 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 1.5 x 0.75 in. | M710, BMP71 | 38.1 mm |
62.75 €
Pack of 250 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||
17828904
|
Brady™
173341 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 1.5 x 1 in. | M710, BMP71 | 38.1 mm |
72.80 €
Pack of 250 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17899094
|
Brady™
174606 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 1.5 x 1.5 in. | M710, BMP71 | 38.1 mm |
90.00 €
Pack of 250 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17808974
|
Brady™
174272 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 1.5 x 0.75 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 38.1 mm |
67.15 €
Pack of 250 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||
17859034
|
Brady™
174075 |
ASTM D 1000 | 1.25 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 31.75 mm |
84.55 €
Pack of 250 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||
17818474
|
Brady™
174237 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 2.5 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 63.5 mm |
45.05 €
Pack of 100 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17898844
|
Brady™
173376 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 2.5 x 1 in. | M710, BMP71 | 63.5 mm |
42.20 €
Pack of 100 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17808444
|
Brady™
173851 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 4 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 101.6 mm |
73.45 €
Pack of 100 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||
17859054
|
Brady™
174607 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 4 x 1.5 in. | M710, BMP71 | 101.6 mm |
68.70 €
Pack of 100 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
17871679
|
Brady™
173768 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 4 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 101.6 mm |
47.60 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17898124
|
Brady™
173860 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 6 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 152.4 mm |
60.90 €
Pack of 50 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||
17838244
|
Brady™
174195 |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | 0.275 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 6.99 mm |
104.00 €
Pack of 750 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
Description
- Adhesive and cloth backing offers great holding power, yet allow for clean removal and repositioning
- Sticks to any clean, dry surface
- Specially formulated topcoat for high print quality
- Semi-gloss finish for crisp, clean look
- Made of B-498 vinyl material
Specifications
Not Autoclavable | |
None | |
Semi-gloss | |
Vinyl | |
Thermal Transfer | |
Non-sterile | |
No | |
-40°C to +80°C | |
0.185 mm |
Repositionable Rubber | |
White | |
Die-cut | |
Single-side Printable | |
Labels | |
Barcode Labeling, Circuit Board Labeling, Component and Equipment Labeling, Laboratory Labeling, Wire and Cable Labeling | |
-40°F to +176°F | |
0.0073 in. |