Learn More
Cytiva Research Grade FBS, Irradiated and Heat Inactivated
Research grade Fetal Bovine Serum of South America Origin. Irradiated and Heat inactivated.
Brand:  Cytiva SV30160.03IH
Product Code. 15745533
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Supplement research cell culture growth with South America-sourced quality HyClone Fetal Bovine Serum.
  • Sourced in EU-approved South American countries with complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Produced using true pool technology for uniformity and consistency from bottle to bottle
  • Filtered through three sequential 100nm (0.1um) pore size-rated filters
  • Virus panel testing according to 9 CFR 113.53
TRUSTED_SUSTAINABILITY
Specifications
Specifications
South America
500 mL
Research grade