Learn More
WTW™ Resistance Test Kit for WTW™ Conductivity Meter
For conductivity measuring systems, there are different check methods:The system including measuring cell can be calibrated with suitable traceably certifed standard solutions. An adjustment is usually not necessary due to the long-term stability of the used electrode materials. Special kits contain all necessary parts to test conductivity measuring systems according to pharamcopeia guidelines.
Brand:  WTW™ 302870
Product Code. 10191721
  • 595.00 € / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Resistance test kit
Waterproof 8-pin socket
Checking all conductivity meters