Learn More
Macherey-Nagel™ Resolvosil BSA-7 CC Guard Columns Cartridges
Brand:  Macherey-Nagel™ 721702.40
Product Code. 10681102
  • 139.00 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1 Pack
8 mm
CC Guard Columns Cartridges
EC columns of 4 and 4.6 mm ID with guard column adapter EC (REF 721359)
7 μm
300Å
4 mm
Resolvosil BSA-7