missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva RESOURCE™ S IEX Columns
RESOURCE prepacked columns for fast separation. Resource S IEX Columns are prepacked with Source 15S, a strong cation exchanger for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.
Brand: Cytiva 17-1180-01
2448.15 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics, sample loading of up to 25mg/mL and stability in organic solvents and pH extremes
- Resource columns with 1mL bed volume have separation times of 3 minutes or less at 9.6mL/min. and approximately 20 min. using a peristaltic pump at 1mL/min.
Specifications
RESOURCE S | |
PEEK | |
>60 mL/min. | |
Sulphonate group | |
16 mm | |
15 μm | |
1 x 6 mL |
Connectors for high- and low-pressure chromatography equipment and instructions | |
Polystyrene/divinylbenzene | |
RESOURCE S columns are prepacked with SOURCE 15S, a strong cation exchanger for high-resolution polishing purification of proteins, at high flow rates. | |
4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol | |
6 mL | |
2 to 13 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- RESOURCE S, 6 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.