Learn More
Thermo Scientific™ Restore™ PLUS Western Blot Stripping Buffer
Remove high-affinity antibodies from membranes so they can be reprobed and detected with chemiluminescent substrates.
60.25 € - 250.00 €
Specifications
Form | Liquid |
---|---|
Product Line | Restore™ |
Content And Storage | Upon receipt store at room temperature. Product shipped at ambient temperature. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11829380
|
Thermo Scientific™
46428 |
30 mL |
60.25 €
30mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10016433
|
Thermo Scientific™
46430 |
500 mL |
250.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific™ Restore PLUS Western Blot Stripping Buffer is an advanced formula for removing bound primary and secondary antibodies from membranes so they can be reprobed and detected with chemiluminescent substrates.
Restore PLUS Buffer is an alternative formulation of the original Thermo Scientific Restore Western Blot Stripping Buffer. Restore PLUS Stripping Buffer was designed for use with antibodies that are difficult to remove from Western blots and require longer incubation times or incubation temperatures greater than 22°C with gentler formulations. High-affinity antibodies can be quickly and effectively stripped from Western blots at room temperature without removing transferred proteins, thereby allowing multiple reprobes of the target.Highlights:
Ready and easy to use – no dilution necessary; no offensive odors; store at room temperature
Compatible – use on nitrocellulose and PVDF membranes, whether still wet or already dry; works with practically any blocking buffer, enzyme conjugate and chemiluminescent substrate
Cost effective – save valuable time and samples; strip blots effectively the first time
Robust yet gentle – transferred proteins remain viable; strip the same blot up to five times
Flexible – strip and reprobe to optimize antibody concentrations or to detect a new antigen with different antibodies
Specifications
Liquid | |
Restore™ | |
Upon receipt store at room temperature. Product shipped at ambient temperature. |
Safety and Handling
- RESTORE PLUS WESTERN BLOT
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
Supplemental information
- MIXTURE LIST-Contains : Anionic Surfactant ANIONIC SURFACT ( 1-3)
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.