missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Resurge™ CD3 Supplement, without glucose and L-glutamine

A diverse set of fully chemically defined, animal origin–free and protein-free cell culture supplements, which help promote cell growth and boost protein production.

166.00 € - 7660.00 €

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Cell Line CHO
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Powder
Product Type Media Supplement
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16259911
View Documents
Gibco™
670019
1 kg
1642.00 €
1kg
Please to purchase this item. Need a web account? Register with us today!
16269911
View Documents
Gibco™
670020
5 kg
7660.00 €
5kg
Please to purchase this item. Need a web account? Register with us today!
16249911
View Documents
Gibco™
670018
100 g
166.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

An animal origin-free (AOF) protein-free chemically defined supplement. It can be used in batch and fed batch mode in cell culture processes. All components are chemically defined with known molecular formulae and structures and meet our stringent Animal Origin-Free (AOF) specification. Formulated without glucose, L-glutamine, and sodium bicarbonate. Due to the balanced nutritional nature of this supplement, rich in amino acids, it has also been used in bacterial applications.
Specifications

Specifications

Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Animal Origin-free, Chemically-defined, Protein-free
Media Supplement
Resurge™
CHO
Powder
Bottle(s)
Animal origin-free
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.