Learn More
Gibco™ Resurge™ CD4 Supplement, without glucose and L-glutamine
A diverse set of fully chemically defined, animal origin–free and protein-free cell culture supplements, which help promote cell growth and boost protein production.
Brand:  Gibco™ 670023
 View more versions of this product
Product Code. 16299911
  • 7660.00 € / 5kg
Explore more special offers
Description
Description
A chemically defined supplement formulation to complement a variety of base media. This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Animal Origin-free, Chemically-defined, Protein-free
Powder
No Glutamine
Resurge™
Animal origin-free
CHO
Mammalian Cell Culture
Media Supplement
Bottle(s)
5 kg