Learn More
Applied Biosystems™ 3500 Dx Series 96-Well Fast (0.1 mL) Retainer & Base Set
This is a 96-well retainer & base set (fast) designed for use with the 3500 Dx and 3500xL Dx genetic analyzers.
Brand:  Applied Biosystems™ 4410229
Product Code. 17296483
  • 333.00 € / Pack of 4
Explore more special offers
Description
Description
This is a retainer and base set (fast) designed for septa retention in 8-tube strip and 96-well plate formats for use with the 3500 Dx and 3500xL Dx genetic analyzers.
Note: The 3500 Dx and 3500xL Dx genetic analyzers and consumables are for distribution and use in select countries only.
  • This is a 96-well retainer & base set (fast) designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer
  • Septa and plate base⁄retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers.
  • The 3500 Dx Series Systems and consumables are for distribution and use in select countries only. Not for use in the USA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
96-Well Microtiter Plates, MicroAmp™ Optical 96-Well Reaction Plates
96-well
Retainer and Base Set
Room Temperature
Ambient storage.
Each package contains 4 sets.
3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
4 Sets
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ 3500 Dx Series 96-Well Fast (0.1 mL) Retainer & Base Set > 4 Sets
For In Vitro Diagnostic Use Only