missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Retinoic acid, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Retinoic acid is an endogenous agonist for retinoic acid receptors (IC50 = 14 nM for RARα, RARβ and RARγ receptors). Also promotes differentiation of mouse embryonic stem cells (ESCs) into adipocytes, neurons and glia in vitro. Proposed ligand of RORβ (Kd = 280 nM). Activates autophagy.
For more information about how Retinoic acid may be used, see our protocols: Generation of β cells from hPSCs, Cultivating Cerebral Organoids .
Specifications
Specifications
CAS | 302-79-4 |
Molecular Formula | C20H28O2 |
MDL Number | MFCD00001551 |
Quantity | 50 mg |
Synonym | retinoic acid, tretinoin, vitamin a acid, all-trans-retinoic acid, trans-retinoic acid, renova, airol, retin-a, vesanoid, atra |
InChI Key | SHGAZHPCJJPHSC-YCNIQYBTSA-N |
SMILES | C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O |
IUPAC Name | (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid |
Molecular Weight (g/mol) | 300.44 |
PubChem CID | 444795 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'