Learn More
Fisherbrand™ Reusable Class A Volumetric Pipets
Available in either standard or serialized models.
Brand:  Fisherbrand™ 13-650-2D
 View more versions of this product
Product Code. 17871440
  • 101.00 € / Pack of 12
Explore more special offers
Description
Description
Available in either standard or serialized models.
  • Borosilicate glass
  • Volume accuracy marked on each pipet
  • Permanent amber markings
  • Tuf-Temp™–treated tempered tips reduce chipping
Order Info
All pipets are available as PersonaPet™ personalized pipets.
Specifications
Specifications
±0.01 mL
ASTM E969-95
Borosilicate Glass
Amber
Tuf-Temp™ treated
Black
Volumetric Pipet
Yes
A
Yes
8 sec.
0.1 oz.
3 mL
Reusable