Learn More
Thermo Scientific™ Reusable Cryo Sleeve
Utilize the Thermo Scientific™ Reusable Cryo Sleeve with cane and canister systems.
Brand: Thermo Scientific™ 4000218
182.40 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
Utilize the Thermo Scientific™ Reusable Cryo Sleeve with cane and canister systems.
Specifications
Polyvinyl Sleeve; I.D. x L: 0.6 in. (1.5cm) x 10.8 in. (27.4cm); Case of 100 | |
Cryo Sleeve | |
CryoPlus 1,2,3,4 |