Learn More
Fisherbrand™ Reverse Osmosis Membrane with Housing
Deionizes tap water and filters out particles for laboratory procedures. Fisher Scientific™ Reverse Osmosis Membrane with Housing is for use with the Accu500 Ultrapure Water System.
Brand:  Fisherbrand™ RAR0100HG
 View more versions of this product
Product Code. 15469669
  • 445.00 € / Each
Estimated Shipment: 12-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Accu500.
  • Two sizes: 10L / 20L or 15L / 30L
  • Removes particles, ions, and organics from tap water
Specifications
Specifications
Accu500 10/20L
Reverse Osmosis Membrane
Housing