Learn More
Grant Instruments™ Reverse Spin Bioreactor for Mixing and Oxygenating Aerobic Cultivation
The Grant Instruments RTS-1 is personal bioreactor which provides 'Reverse-Spin' type of agitation and logging of microbial growth in 50 mL tubes in real time. RTS-1 including TubeSpin™ Bioreactor 50, TPP™, 20 pcs.
Brand:  Grant Instruments™ RTS-1 EU
Product Code. 17715029
  • 4365.00 € / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Features:
  • Innovative mixing due to reverse spinning of the sample around its own axis
  • Due to innovative mixing technology it is possible to measure optical density of the probe in real time
  • Store, display and analyse the data in real time
  • Compact device with low profile and small footprint for personal application
  • Temperature control allows use as an incubator, e.g. for cell growth
  • The ability to change parameters such as temperature, RPM and “Reverse-Spin” frequency, allows to achieve consistency and reproducibility of results
  • Programmable Cycling/profiling of cultivation parameters such as temperature, RPM, “Reverse-Spin” frequency
  • The ability to remotely monitor the process of cultivation while home or using a mobile phone
Software features:
  • Real–Time cell growth logging
  • 3D graphical representation of OD or growth rate over time over unit
  • Pause option
  • Save/Load option
  • Report option: PDF and Excel
  • Connect up to 12 units simultaneously to 1 computer
  • Remote monitoring option (requires internet connection)
  • Cycling/Profiling options
  • User calibration options for any microorganism
Applications:
  • Bacterial cultivation with real time growth kinetics
  • Strain screening
  • Temperature stress and fluctuation experiments
  • Media screening and optimization
  • Synthetic and systems biology
  • Inhibition and toxicity tests
Specifications
Specifications
EU plug