missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Reversible Microtube Racks with Lid

Product Code. 11771253 Shop All MBP Molecular BioProducts Products
Click to view available options
Color:
Blue
Blue, Green, Red, Yellow
Fluorescent Green
Fluorescent Green, Fluorescent Orange, Fluorescent Pink, Purple
Fluorescent Orange
Green
Pink
Red
Yellow
Unit Size:
Each
Pack of 12
This item is not returnable. View return policy

Product Code. 11771253

Brand: Thermo Scientific™ 863011

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Store samples securely and rapidly identify them with this reversible microtube rack with lid, which features a low-profile design with an alphanumeric grid.

Store samples securely and rapidly identify them with Thermo Scientific™ Reversible Microtube Racks with Lid. These racks feature a stable, low-profile design with an alphanumeric grid.

  • Hold 0.2mL tubes on one side, 0.5mL tubes on the other
  • Lid protects samples and allows stacking
  • 100% Recycled plastic
  • Several colors available
TRUSTED_SUSTAINABILITY

Specifications

Product Type Reversible Microtube Rack
Description Reversible Microtube Rack
Color Blue
Holds Tubes (0.2 mL), Tubes (0.5 mL)
Material Recycled Plastic
Stackable Yes
No. per Case 12 Racks
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.