Learn More
Grant Instruments™ Reversible Raised Shelf for Unstirred Water Baths
Allows two shelf depths to accommodates shallow vessels. Grant Instruments™ Reversible Raised Shelf for Unstirred Water Baths can be used with a variety of other Grant Heating Circulators and Heated Circulating Baths.
Brand:  Grant Instruments™ RS14H
 View more versions of this product
Product Code. 11446407
  • 179.00 € / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Choose from a variety of sizes to fit your tank.
  • For use with either Stainless Steel and Plastic tank models
Specifications
Specifications
Half the area of SBB Aqua 12 Plus, Sub Aqua 12 Pro, or Sub Aqua Pro Dual
Stainless Steel
40 or 78 mm