Learn More
RGX-104, MedChemExpress
MedChemExpress RGX-104 is an orally bioavailable and potent liver-X nuclear hormone receptor (LXR) agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
Brand:  Medchem Express HY-111498A
Encompass_Preferred
 View more versions of this product
Product Code. 16444478
  • 359.10 € / 10mg
Explore more special offers
Description
Description
RGX-104 is an orally bioavailable and potent liver-X nuclear hormone receptor (LXR) agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
Specifications
Specifications
RGX-104
White
C34H33ClF3NO3
DMSO : 100 mg/mL (167.76 mM; Need ultrasonic)
596.08
99.35%
Solid
610318-54-2
Research
10 mg
O=C(O)CC1=CC=CC(OCC[C@H](N(CC2=CC=CC(C(F)(F)F)=C2Cl)CC(C3=CC=CC=C3)C4=CC=CC=C4)C)=C1
596.08
Research
Cancer-programmed cell death