Learn More
RGX-104, MedChemExpress
MedChemExpress RGX-104 is an orally bioavailable and potent liver-X nuclear hormone receptor (LXR) agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
Brand: Medchem Express HY-111498A
This item is not returnable.
View return policy
Description
RGX-104 is an orally bioavailable and potent liver-X nuclear hormone receptor (LXR) agonist that modulates innate immunity via transcriptional activation of the ApoE gene.Specifications
RGX-104 | |
White | |
C34H33ClF3NO3 | |
DMSO : 100 mg/mL (167.76 mM; Need ultrasonic) | |
596.08 | |
99.35% | |
Solid |
610318-54-2 | |
Research | |
100 mg | |
O=C(O)CC1=CC=CC(OCC[C@H](N(CC2=CC=CC(C(F)(F)F)=C2Cl)CC(C3=CC=CC=C3)C4=CC=CC=C4)C)=C1 | |
596.08 | |
Research | |
Cancer-programmed cell death |