Learn More
Rotronic™ Rh Probe Hk25 Temperature
Product Code. 12849387
Product Code. 12849387
Brand: Rotronic™ SPM15FS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
humidity, moisture in ducts, dryers, chambers,ovens polycarbonate for HP22 250mm x 15mm (l x dia.) -100 to +150°C, 0 to 100%RH HygroClip2