missing translation for 'onlineSavingsMsg'
Learn More
RHC 80267, Tocris Bioscience™ Product Code.: 18046062

RHC 80267, Tocris Bioscience™

Product Code. 18046062
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18046062

Brand: Tocris Bioscience 1842/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Diacylglycerol lipase inhibitor

Inhibitor of diacylglycerol lipase (IC50 values are 1.1 and 4 μM in rat cardiac myocytes and canine platelets respectively). Weakly inhibits phospholipases C and A2. Potentiates acetylcholine evoked relaxation in mesenteric arteries by the inhibition of cholinesterase activity (IC50 = 4 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83654-05-1
Molecular Formula C20H34N4O4
Molecular Weight (g/mol) 394.52
MDL Number MFCD00210844
InChI Key RXSVYGIGWRDVQC-UHFFFAOYSA-N
Synonym 1,6-bis cyclohexyloximinocarbonyl hexane, 1,6-di o-carbamoyl cyclohexanone oxime hexane, 1,6-bis cyclohexyloximinocarbonylamino hexane, cyclohexanone, o,o'-1,6-hexanediylbis iminocarbonyl dioxime, quinolyl derivatives, d05lxg, cyclohexanone, o,o'-1,6-hexanediylbis iminocarbonyl oxime, rhc hplc , solid, o,o'-1,6-hexanediylbis iminocarbonyl dioxime cyclohexanone, cyclohexylideneamino n-6-cyclohexylideneamino oxycarbonylamino hexyl carbamate
PubChem CID 5063
IUPAC Name cyclohexylideneamino N-[6-({[(cyclohexylideneamino)oxy]carbonyl}amino)hexyl]carbamate
SMILES O=C(NCCCCCCNC(=O)ON=C1CCCCC1)ON=C1CCCCC1

Specifications

Quantity 10 mg
Formula Weight 394.51
Percent Purity >95%
Chemical Name or Material RHC 80267
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
RHC 80267, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.