Learn More
Thermo Scientific™ RheFlex™ High-Pressure Fittings
Ensure reliable high-pressure connections with Thermo Scientific™ RheFlex™ High-Pressure Fittings.
24.00 € - 216.12 €
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
10016844
|
Thermo Scientific™
6000-109 |
Short High Pressure Fittings Set | 5/Pk. | N/A | |||||
10650816
|
Thermo Scientific™
6000-110 |
High Pressure 1/16 in. Ferrule | 5/Pk. | N/A | |||||
10249704
|
Thermo Scientific™
6000-111 |
Long High Pressure Fittings Set | 5/Pk. | N/A | |||||
11567701
|
Thermo Scientific™
6000-162 |
Extra Long High Pressure Fittings Set | 5/Pk. | N/A | |||||
11577701
|
Thermo Scientific™
6000-209 |
Short High Pressure Fittings Set | 10/Pk. | N/A | |||||
10299594
|
Thermo Scientific™
6000-210 |
High Pressure 1/16 in. Ferrule | 10/Pk. |
57.90 €
Pack of 10 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
||||
10269264
|
Thermo Scientific™
6000-211 |
Long High Pressure Fittings Set | 10/Pk. | N/A | |||||
10197704
|
Thermo Scientific™
6000-262 |
Extra Long High Pressure Fittings Set | 10/Pk. |
216.12 €
Pack of 10 |
Estimated Shipment: 30-01-2025
Log in to see stock. |
||||
Description
Ensure reliable high-pressure connections by using Thermo Scientific™ RheFlex™ High-Pressure Fittings. These fittings are precision machined from 316 stainless steel to be burr- and contaminant-free.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.