missing translation for 'onlineSavingsMsg'
Learn More

Rhenium, 1% on 2.5mm alumina spheres, Thermo Scientific™

Product Code. 15472827
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-15-5
Physical Form Spheres
Quantity 250 g
Molecular Formula Re
MDL Number MFCD00011195
Synonym rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
Solubility Information Insoluble in water.
InChI Key WUAPFZMCVAUBPE-UHFFFAOYSA-N
SMILES [Re]
IUPAC Name rhenium
Molecular Weight (g/mol) 186.21
PubChem CID 23947
ChEBI CHEBI:49882
Chemical Name or Material Rhenium, 1% on alumina spheres, 2.5mm alumina spheres
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'