missing translation for 'onlineSavingsMsg'
Learn More

Rhenium foil, 0.1mm (0.004in) thick, 99.97% (metals basis)

Product Code. p-7048372
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
This item is not returnable. View return policy

Product Code. 11301408

Brand: Thermo Scientific Alfa Aesar 010308.FF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-15-5 | Re | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-15-5
Molecular Formula Re
Molecular Weight (g/mol) 186.21
MDL Number MFCD00011195
InChI Key WUAPFZMCVAUBPE-UHFFFAOYSA-N
Synonym rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
PubChem CID 23947
ChEBI CHEBI:49882
IUPAC Name rhenium
SMILES [Re]

Specifications

Quantity 25 x 25 mm
Odor Odorless
Weight ≈1.32g/25x25mm
Form Foil
Percent Purity 99.97%
Assay (metals basis)
Chemical Name or Material Rhenium foil, 0.1mm (0.004 in.) thick

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.