missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhenium(III) chloride, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.25 g
1 g
Description
It is used as a starting material for synthesis of rhenium complexes. ReCl3 can react with peralkylated benzene-metal complexes that act as electron reservoirs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Red |
Physical Form | Powder |
Quantity | 0.25 g |
Solubility Information | Soluble in water. |
Formula Weight | 292.56 |
Odor | Odorless |
Sensitivity | Moisture sensitive |
Appearance | Dark red powder |
Chemical Name or Material | Rhenium(III) chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'