missing translation for 'onlineSavingsMsg'
Learn More

Rhenium powder, 99.99%, Thermo Scientific Chemicals

Product Code. 11311418
Click to view available options
Quantity:
0.2 g
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7440-15-5
Molecular Formula Re
Molecular Weight (g/mol) 186.21
MDL Number MFCD00011195
InChI Key WUAPFZMCVAUBPE-UHFFFAOYSA-N
PubChem CID 23947
ChEBI CHEBI:49882
IUPAC Name rhenium
SMILES [Re]

Especificaciones

Appearance Powder
Assay Percent Range 99.99% (Metals basis)
Quantity 0.2 g
Synonym rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
Mesh Size -325 Mesh
Form Powder
Chemical Name or Material Rhenium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'