missing translation for 'onlineSavingsMsg'
Learn More

Rhenium(VI) oxide, 99.9% (metals basis)

Product Code. 11314779
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11314779

Brand: Thermo Scientific Alfa Aesar 062109.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rhenium(VI) oxide, O3Re, CAS Number-1314-28-9, rhenium vi oxide trace metals basis 1g, rhenium vi oxide, rhenium trioxide, rhenia, rhenium oxide reo3, reo3, unii-fet0y2c413, 5g, 99.9% (metals basis), 400 deg.C (decomposition), 215-228-8, 236.2, YSZJKUDBYALHQE-UHFFFAOYSA-N, trioxorhenium | CAS: 1314-28-9 | O3Re | 234.204 g/mol

Rhenium(VI) oxide is involved as a catalyst for the hydrogenation of carboxylic acids and carboxamides. It is involved in the alkene epoxidation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 1314-28-9
Molecular Formula O3Re
Molecular Weight (g/mol) 234.204
MDL Number MFCD00016306
InChI Key YSZJKUDBYALHQE-UHFFFAOYSA-N
Synonym rhenium trioxide, rhenium oxide reo3, rhenium vi oxide, unii-fet0y2c413, rhenia, reo3, rhenium vi oxide trace metals basis 1g
PubChem CID 102110
IUPAC Name trioxorhenium
SMILES O=[Re](=O)=O

Specifica

Boiling Point 400°C (decomposition)
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 236.2
Sensitivity Moisture sensitive
Chemical Name or Material Rhenium(VI) oxide

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato