Learn More
Rhenium(VII) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Moisture Sensitive; Sublimation temperature/start: 250°C (482°F)
Chemical Identifiers
CAS | 1314-68-7 |
---|---|
Molecular Formula | O7Re2 |
Molecular Weight (g/mol) | 484.407 |
MDL Number | MFCD00011199 |
InChI Key | NBGOSNNAAHDRLK-UHFFFAOYSA-N |
Synonym | rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basis |
PubChem CID | 123106 |
IUPAC Name | trioxo(trioxorheniooxy)rhenium |
SMILES | O=[Re](=O)(=O)O[Re](=O)(=O)=O |
Description
Rhenium(VII) oxide acts as a precursor to methylrhenium trioxide, organorhenium oxides which catalyze olefin oxidation, metathesis and other transformations. It is used as a reagent for dehydrations and rearrangements of allylic alcohols, oximes and amides, oxidative cyclizations, alkene metathesis and formation of bisperoxyacetals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-68-7 | |
484.407 | |
NBGOSNNAAHDRLK-UHFFFAOYSA-N | |
123106 | |
O=[Re](=O)(=O)O[Re](=O)(=O)=O |
O7Re2 | |
MFCD00011199 | |
rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basis | |
trioxo(trioxorheniooxy)rhenium |
Specifications
360°C | |
1314-68-7 | |
Crystalline | |
O7Re2 | |
rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basis | |
NBGOSNNAAHDRLK-UHFFFAOYSA-N | |
trioxo(trioxorheniooxy)rhenium | |
123106 | |
Odorless | |
Yellow powder | |
Rhenium(VII) oxide |
297°C | |
White | |
99.99% (metals basis) | |
MFCD00011199 | |
Soluble in water and alcohol. | |
O=[Re](=O)(=O)O[Re](=O)(=O)=O | |
484.407 | |
484.4 | |
Moisture sensitive | |
6.103 g/cm3 at 20°C |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: III
missing translation for 'einecsNumber' : 215-241-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only