Learn More
Thermo Scientific™ Rheodyne™ 7010 Sample Injector
Ensure accurate and precise high-volume HPLC injections with the single-mode Thermo Scientific™ Rheodyne™ 7010 Sample Injector.
Brand:  Thermo Scientific™ 7010
Product Code. 10008952
  • 1479.68 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensure accurate and precise high-volume HPLC injections of 5μL to 20mL samples with the Thermo Scientific™ Rheodyne™ 7010 Sample Injector. This single-mode injector uses the complete filling method to load the sample loop.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Single Mode
Sample Injector
Stainless Steel
5 μL to 5 mL
6 Ports