missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Rhodamine 101 inner salt, 94%, pure
Click to view available options
Quantity:
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 271.0°C |
CAS | 116450-56-7 |
Assay Percent Range | 99% |
Molecular Formula | C32H30N2O3 |
Fluorescence Max | 588nm (in methanol) |
MDL Number | MFCD00051383 |
InChI Key | MUSLHCJRTRQOSP-UHFFFAOYSA-N |
Infrared Spectrum | Authentic |
SMILES | C1CC2=C3C(=C4C(=C2)C(=C5C=C6CCC[N+]7=C6C(=C5O4)CCC7)C8=CC=CC=C8C(=O)[O-])CCCN3C1 |
Molecular Weight (g/mol) | 490.603 |
Show More |
Safety and Handling

Product Identifier
- Rhodamine 101 inner salt
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap and water.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/physician/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals Rhodamine 101 inner salt, 94%, pure >
Spot an opportunity for improvement?Share a Content Correction