missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Rhodamine 6G, 99%, pure, Thermo Scientific Chemicals
51.86 € - 275.37 €
Chemical Identifiers
CAS | 989-38-8 |
---|---|
Molecular Formula | C28H31ClN2O3 |
Molecular Weight (g/mol) | 479.017 |
MDL Number | MFCD00012665 |
InChI Key | VYXSBFYARXAAKO-UHFFFAOYSA-N |
Synonym | C.I. 45160, Basic Red 1 |
PubChem CID | 51358423 |
IUPAC Name | ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride |
SMILES | [H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10732792
|
Thermo Scientific Chemicals
419025000 |
500 mg | N/A | ||||||
10657381
|
Thermo Scientific Chemicals
419020010 |
1 g | N/A | ||||||
11956931
|
Thermo Scientific Chemicals
419020050 |
5 g |
275.37 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
989-38-8 | |
479.017 | |
VYXSBFYARXAAKO-UHFFFAOYSA-N | |
51358423 | |
[H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-] |
C28H31ClN2O3 | |
MFCD00012665 | |
C.I. 45160, Basic Red 1 | |
ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride |
Specifications
290.0°C | |
99% | |
MFCD00012665 | |
C.I. 45160, Basic Red 1 | |
VYXSBFYARXAAKO-UHFFFAOYSA-N | |
ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride | |
51358423 | |
99% | |
Glass bottle |
989-38-8 | |
C28H31ClN2O3 | |
18, V,12, 283 | |
Solubility in water: soluble. | |
[H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-] | |
479.017 | |
479.01 | |
Pure | |
Rhodamine 6G |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Rhodamine 6G
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title