missing translation for 'onlineSavingsMsg'
Learn More
Rhodanine 99%, Thermo Scientific Chemicals Product Code.: 10582931

Rhodanine 99%, Thermo Scientific Chemicals

Product Code. 10582931
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10582931

Brand: Thermo Scientific Acros 152600250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-84-4
Molecular Formula C3H3NOS2
Molecular Weight (g/mol) 133.183
MDL Number MFCD00005488
InChI Key KIWUVOGUEXMXSV-UHFFFAOYSA-N
Synonym 2-Thioxo-4-thiazolidinone
PubChem CID 1201546
ChEBI CHEBI:8830
IUPAC Name 2-sulfanylidene-1,3-thiazolidin-4-one
SMILES C1C(=O)NC(=S)S1

Specifications

Melting Point 167°C to 170°C
Assay Percent Range 99%
Beilstein 27, 242
Merck Index 15, 8308
Infrared Spectrum Authentic
Formula Weight 133.2
Percent Purity ≥98.5%
Packaging Glass bottle
Color Beige to Yellow
Quantity 25 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Rhodanine, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0439795 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number152601000,15260037,152600250
Lot NumberA0459329 Certificate TypeCertificate of Analysis Date15/01/2024 Catalog Number152601000,15260037,152600250
Lot NumberA0395394 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number152601000,15260037,152600250
Lot NumberA0405276 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number152601000,15260037,152600250
Lot NumberA0447317 Certificate TypeCertificate of Analysis Date09/11/2022 Catalog Number152601000,15260037,152600250
compliance-icons
Product Identifier
  • Rhodanine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhodanine 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.