missing translation for 'onlineSavingsMsg'
Learn More

Rhodium, 5% on alumina powder, reduced, Thermo Scientific™

Product Code. 11311838
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]

Specifications

Color Black
Appearance Black powder
Quantity 2 g
Solubility Information Insoluble in water.
Form Powder
Chemical Name or Material Rhodium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'