missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(II) acetate, dimer, 98+%, Thermo Scientific Chemicals

Product Code. 11358176
Click to view available options
Quantity:
100 mg
500 mg
2 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Catalyst for cyclopropanation of alkenes, oxidation of alcohols, cyclization reactions involving ?-diazo carbonyl groups, insertion into C-H and X-H bonds (X-H is NH or SH or OH) and for ylide formation. More reactive and useful in differentiating ribonucleosides and deoxynucleosides. It is also used in functionalizing fullerenes into polymers. Efficient catalyst for hydrogen transfer from 2-propanol to cyclohexanone and other unsaturated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Catalyst for cyclopropanation of alkenes, oxidation of alcohols, cyclization reactions involving ɑ-diazo carbonyl groups, insertion into C-H and X-H bonds (X-H is NH or SH or OH) and for ylide formation. More reactive and useful in differentiating ribonucleosides and deoxynucleosides. It is also used in functionalizing fullerenes into polymers. Efficient catalyst for hydrogen transfer from 2-propanol to cyclohexanone and other unsaturated compounds.

Solubility
Soluble in aqueous solution like water. Slightly soluble in ethanol

Notes
Stable under normal conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15956-28-2
Molecular Formula C8H12O8Rh2
Molecular Weight (g/mol) 441.99
MDL Number MFCD00003538
InChI Key VOXKIADTKKPBBP-UHFFFAOYSA-N
Synonym rhodium ii acetate, rhodium diacetate, acetic acid, rhodium 2+ salt, rhodium ii acetate dimer dihydrate, dirhodium tetraacetate, rhodium ii acetate dimer, rhodium acetate, brown, water soluble, diacetoxyrhodium
PubChem CID 122130469
SMILES C[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4

Specifications

Melting Point 205°C
Quantity 100 mg
Linear Formula [Rh(CO2CH3)2]2
Solubility Information Soluble in aqueous solution like water. Slightly soluble in ethanol
IUPAC Name bis(λ²-rhodium(2+)) tetraacetate
Formula Weight 441.99
Percent Purity ≥98%
Odor Odorless
Chemical Name or Material Rhodium(II) acetate, dimer

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'